Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMOD2 (B01P), Mouse anti-Human, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002843 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-843 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-843 Supplier Abnova Corporation Supplier No. H00029767B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TMOD2 protein.

This gene encodes a neuronal-specific member of the tropomodulin family of actin-regulatory proteins. The encoded protein caps the pointed end of actin filaments preventing both elongation and depolymerization. The capping activity of this protein is dependent on its association with tropomyosin. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq

Sequence: MALPFQKELEKYKNIDEDELLGKLSEEELKQLENVLDDLDPESAMLPAGFRQKDQTQKAATGPFDREHLLMYLEKEALEQKDREDFVPFTGEKKGRVFIPKEKPIETRKEEKVTLDPELEEALASASDTELYDLAAVLGVHNLLNNPKFDEETANNKGGKGPVRNVVKGEKVKPVFEEPPNPTNVEISLQQMKANDPSLQEVNLNNIKNIPIPTLREFAKALETNTHVKKFSLAATRSNDPVAIAFADMLKVNKTLTSLNIESNFITGTGILALVEALKENDTLTEIKIDNQRQQLGTAVEMEIAQMLEENSRILKFGYQFTKQGPRTRVAAAITKNNDLVRKKRVEADRR

Specifications

Antigen TMOD2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TMOD2 protein.
Formulation PBS with no preservative; pH 7.4
Gene TMOD2
Gene Accession No. NM_014548
Gene Alias MGC39481/N-TMOD/NTMOD
Gene Symbols TMOD2
Host Species Mouse
Immunogen TMOD2 (NP_055363.1, 1 a.a. ∼ 351 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29767
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.