Learn More
Description
Tumor necrosis factor α (TNF-α) is a macrophage/monocyte cytokine, usually found as a soluble secreted protein, which has diverse immunomodulatory, anti-tumor, and toxic effects.
Host Species: Mouse
Clone: J1D9
Isotype: IgG1
Cross Reactivity: Human; does not react with mouse. Others-not known.
Immunogen: Recombinant human TNF-α
Molecular Weight: 17kDa
Positive Control: TNF-α recombinant protein
Cellular Localization: Cytoplasmic and extracellular
Ab-1 neutralizes the recombinant TNF-alpha mediated cytotoxicity of L929 cells and inhibits the tumor growth in mice. It protects mice against the toxicity of recombinant human TNF-alpha.
Cytotoxicity Assay, ELISA, Flow Cytometry, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | TNF alpha Ab-1 |
| Classification | Monoclonal |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human TNF-α |
| Quantity | 500 μL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.