Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TNF-alpha, Polyclonal

Catalog No. 6542501MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
65-425-01MG 1 mg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-425-01MG Supplier MilliporeSigma™ Supplier No. 6542501MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects TNF-α Clone: in Human, Mouse samples, and it is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Neutralization, Radioimmunoassay, Immuno Blotting

Human Tumor Necrosis Factorα (TNF-α, cachectin) is a cytokine, produced by variety of cell types, that is involved in mediation of the inflammatory response, resistance to infection, and resistance to tumor growth. Human and mouse TNF-α are approximately 79% homologous at the amino acid level and exhibit cross-species reactivity. This antibody recognizes both recombinant and natural human TNF-α and exhibits some (∽30%) cross-reactivity with mouse TNF-α. It exhibits only minor (<1%) cross-reactivity with human TNF-β (lymphotoxin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF-α
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Neutralization, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS.
Host Species Rabbit
Immunogen recombinant, human TNF-α
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes recombinant and monocyte-derived TNF-α. Exhibits some cross-reactivity with lymphotoxin (TNF-β) and mouse TNF-α in L929 cells.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.