Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Mouse anti-Human, APC, Clone: MAB11, BD

Catalog No. BDB551384 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551384 50 Tests
BDB554514 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB551384 Supplier BD Biosciences Supplier No. 551384
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MAb11 antibody reacts with human tumor necrosis factor (TNF, formerly known as TNF-α) protein. TNF is an efficient paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The use of the MAb11 antibody has been reported to cross-react with TNF of rhesus monkey.

Host Species: Mouse
Clone: MAb11
Isotype: IgG1 κ
Species Reactivity: Non-human primate
Immunogen: Recombinant Human TNF

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Applications Flow Cytometry
Classification Monoclonal
Clone MAB11
Conjugate APC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human TNF
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.