Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Human anti-Mouse/Rat, Biotin, Clone: 516D1A1, BD

Catalog No. BDB558415 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558415 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558415 Supplier BD Biosciences Supplier No. 558415
Add to Cart
Add to Cart

Human Monoclonal Antibody

The 516D1A1 Fab antibody reacts with mouse and rat tumor-necrosis factor (TNF, formerly known as TNF-α) proteins. The 516D1A1 Fab was selected by screening a
human Fab phage display library (Hoet et al.) licensed from Dyax Corp (Cambridge, MA) with recombinant mouse TNF protein. The biology and function of the TNF protein has been extensively reviewed (Hehlgans et al.) in the literature.

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Applications ELISA
Classification Monoclonal
Clone 516D1A1
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Human
Immunogen Mouse TNF
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Ig κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.