Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Rat anti-Mouse, Biotin, Clone: MP6-XT3, BD

Catalog No. BDB554415 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554415 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554415 Supplier BD Biosciences Supplier No. 554415
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MP6-XT3 antibody reacts with mouse TNF (TNF-α). Cross reactivity with lymphotoxin-α (LT-α TNF-β) has not been determined.

Host Species: Rat
Clone: MP6-XT3
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse TNF Recombinant Protein

ELISA, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Applications ELISA
Classification Monoclonal
Clone MP6-XT3
Concentration 0.5mg/mL
Conjugate Biotin
Description Tumor Necrosis Factor
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse TNF Recombinant Protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.