Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Rat anti-Mouse/Rat, Unlabeled, Clone: G281-2626, BD

Catalog No. BDB551225 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551225 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551225 Supplier BD Biosciences Supplier No. 551225
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The G281-2626 antibody reacts with mouse and rat tumor necrosis factor (TNF). The immunogen used to generate the G281-2626 hybridoma was recombinant mouse TNF. This is a neutralizing antibody. Cross reactivity with LT-α has not been determined.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Applications ELISA
Classification Monoclonal
Clone G281-2626
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant Mouse TNF
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.