Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Mouse anti-Human, Unlabeled, Clone: MAB1, BD

Catalog No. BDB551220 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551220 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551220 Supplier BD Biosciences Supplier No. 551220
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MAb1 antibody reacts with human tumor necrosis factor (TNF, aka TNF-α) protein. The immunogen used to generate the MAb1 hybridoma was recombinant human TNF. The use of the MAb1 antibody for epitope mapping of human TNF has been described.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Applications ELISA
Classification Monoclonal
Clone MAB1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human TNF
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.