Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Mouse anti-Human, NA/LE, Unlabeled, Clone: MABTNF-A5, BD

Catalog No. BDB552467 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552467 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552467 Supplier BD Biosciences Supplier No. 552467
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MABTNF-A5 antibody, formerly known as 5N, reacts with human tumor necrosis factor, TNF (aka, TNF-α and TNFSF2). TNF is a multifunctional cytokine involved in a variety of immune and inflammatory responses including hemorrhagic necrosis of tumors, septic shock, fever and autoimmune diseases. TNF can regulate the growth and differentiation of many different cell types. It is produced by different activated cell types including macrophages, T lymphocytes, NK cells, dendritic cells, endothelial cells, peripheral blood leukocytes, osteoblasts, astrocytes, mast cells, Kupffer cells, smooth muscle cells, fibroblasts and certain tumor cells. TNF exists in two biologically active forms, i.e., transmembrane and soluble forms. Upon activation, cells express transmembrane TNF glycoproteins that associate as homotrimeric complexes. After enzymatic cleavage, the extracellular region of membrane TNF sheds as a soluble homotrimer. The biologically active form of TNF has been reported to be a trimer. The immunogen used to generate the MABTNF-A5 hybridoma was purified recombinant human TNF. This is an extraordinarily potent neutralizing antibody that is reportedly also useful for ELISA measurements of human TNF.

Host Species: Mouse
Clone: MABTNF-A5
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Recombinant human TNF protein

Functional Assay, Neutralization

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Applications Neutralization
Classification Monoclonal
Clone MABTNF-A5
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Immunogen Recombinant human TNF protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.