Learn More
TNF Rat anti-Mouse, Unlabeled, Clone: MP6-XT22, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559064
Description
The MP6-XT22 antibody reacts with mouse tumor necrosis factor (TNF, also known as TNF-α). The immunogen used to generate this hybridoma was recombinant mouse TNF. This antibody is routinely tested by immunocytochemistry. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Immunocytochemistry

Specifications
TNF | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant mouse TNF | |
0.25 mg | |
Immunology | |
Mouse | |
IgG1 |
Immunocytochemistry | |
MP6-XT22 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.