Learn More
BioLegend™ anti-TNF-α, Clone: MP6-XT22
Description
Host Species: Rat
Clone: MP6-XT22
Isotype: IgG1, κ
Cross Reactivity: Mouse
Immunogen: E. coli-epressed, recombinant mouse TNF-α
Flow Cytometry
Specifications
Specifications
| Antigen | TNF alpha |
| Classification | Monoclonal |
| Clone | MP6-XT22 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Host Species | Rat |
| Immunogen | E. coli-epressed, recombinant mouse TNF-α |
| Purification Method | Unpurified |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.