Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFα Rat anti-Mouse, Clone: MPG-XT3, MilliporeSigma™

Catalog No. 05168MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-168-MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-168-MI Supplier MilliporeSigma Supplier No. 05168
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-TNFalpha Antibody, clone MPG-XT3 is validated for use in NEUT for the detection of TNFα.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFα
Applications Neutralization
Classification Monoclonal
Clone MPG-XT3
Conjugate Unconjugated
Formulation 0.01M Tris-glycine, 0.15M NaCl, pH 7.4
Gene Accession No. NM_000594.2
Gene Symbols TNF, TNFSF2, Cachectin, cachectin, TNF-alpha, DIF, OTTHUMP00000037669, TNFA, TNF-a
Host Species Rat
Immunogen Recombinant murine TNF-α
Purification Method DEAE chromatography
Quantity 1 mg
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.