Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Hamster anti-Mouse/Rat, Unlabeled, Clone: TN3-19.12, BD

Catalog No. BDB557516 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557516 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557516 Supplier BD Biosciences Supplier No. 557516
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The TN3-19.12 antibody reacts with rat and mouse tumor-necrosis factor (TNF) proteins (also known as TNF-α) . Moreover, the TN3-19.12 antibody is reported to crossreact with rabbit TNF, but it does not crossreact with mouse lymphotoxin-α (LT-α, also known as TNF-β) nor with human TNF. The immunogen used to generate the TN3-19.12 hybridoma was E. coli-expressed, purified recombinant mouse TNF protein. This monoclonal antibody has been reported to neutralize the bioactivities of mouse, rat and rabbit TNF. This antibody is routinely tested by ELISA. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Applications ELISA
Classification Monoclonal
Clone TN3-19.12
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Recombinant mouse TNF protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 λ1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.