Learn More
TNF Mouse anti-Human, FITC, Clone: MAB11, BD
Mouse Monoclonal Antibody
$144.60 - $294.00
Specifications
Antigen | TNF |
---|---|
Clone | MAB11 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB554512
|
BD Biosciences
554512 |
0.1 mg |
Each for $294.00
|
|
|||||
BDB552889
|
BD Biosciences
552889 |
50 Tests |
Each for $144.60
|
|
|||||
Description
The MAb11 antibody reacts with human tumor necrosis factor (TNF, formerly known as TNF-α) protein. TNF is an efficient paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The use of the MAb11 antibody has been reported to cross-react with TNF of rhesus monkey.
Host Species: Mouse
Clone: MAb11
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Recombinant Human TNF
Flow Cytometry, Intracellular Staining
Specifications
TNF | |
Flow Cytometry | |
FITC | |
RUO | |
Recombinant Human TNF | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MAB11 | |
Monoclonal | |
Mouse | |
Immunology | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.