Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-TNF p55, Polyclonal, Novus Biologicals™

Supplier: Novus Biologicals NB600627
Specifications
TNF p55 | |
Polyclonal | |
Western Blot 1:500-1:2000, ELISA 1:1000-1:5000 | |
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
In general, this antibody also detects primate sTNFRp55. The antibody does not recognize human sTNFRp75. In ELISA formats and other immunoreactive assays, this antibody will detect the extracellular domain of the TNFRp55 found in human body fluids, particularly in the circulation, urine and supernatants of cells and synovial fluid. This antibody can be used for analysis. Caution should be exhibited as the F (c) domain of the rabbit IgG molecule may interact with cells non-specifically. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
TNFRSF1A | |
The whole rabbit serum used to produce this IgG fraction antibody was prepared by repeated immunizations with recombinant human TNF p55 expressed in Chinese Hamster Cells. | |
500 μL | |
Apoptosis, Asthma, Cancer, Cytokine Research, Diabetes Research, Immunology, Innate Immunity | |
7132 | |
Human, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction