Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TNF RII/TNFRSF1B, Fluorescein, Clone: 22235, R&D Systems™

Catalog No. FAB226F
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB226F 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB226F Supplier R&D Systems Supplier No. FAB226F
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TNF RII/TNFRSF1B
Applications Flow Cytometry
Classification Monoclonal
Clone 22235
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Gene Symbols TNFRSF1B
Host Species Mouse
Immunogen E. coli-derived recombinant human TNF RII Extracellular domain
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7133
Test Specificity Detects human TNF RII in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhLymophotactin beta R, rhNGF R, rhOPG, recombinant mouse (rm) OX40, rhRANK, rhTAJ, rhTNF RI, or rmTNF RII.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.