Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tumor necrosis factor, alpha-induced protein 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004347 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-347 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-347 Supplier Abnova Corporation Supplier No. H00007127A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TNFAIP2.

This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. The expression of this gene was shown to be induced by retinoic acid in a cell line expressing a oncogenic version of the retinoic acid receptor alpha fusion protein, which suggested that this gene may be a retinoic acid target gene in acute promyelocytic leukemia. [provided by RefSeq

Sequence: YILANADTIQHFCTQHGSPATWLQPALPTLAEIIRLQDPSAIKIEVATYATCYPDFSKGHLSAILAIKGNLSNSEVKRIRSILDVSMGAQEPSRPLFSL

Specifications

Antigen tumor necrosis factor, alpha-induced protein 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TNFAIP2.
Formulation 50% glycerol
Gene TNFAIP2
Gene Accession No. NM_006291
Gene Alias B94
Gene Symbols TNFAIP2
Host Species Mouse
Immunogen TNFAIP2 (NP_006282, 552 a.a. ∼ 650 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7127
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.