Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFAIP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA534456
Description
A suggested positive control is K562 cell lysate. PA5-34456 can be used with blocking peptide PEP-1498.
TNFAIP2, also known as B94, is a TNF-a-regulated gene that is expressed in endothelial cells, peripheral blood cells, and mature sperm. Recently, elevated TNFAIP2 expression was observed in nasopharyngeal carcinoma, and high expression of TNFAIP2 was significantly correlated with higher levels of cell migration, invasion and metastasis, suggesting that TNFAIP2 may serve as a useful prognostic indicator for nasopharyngeal carcinoma. TNFAIP2 has also been implicated as part of the viral-sensing circuit of the innate immune response.
Specifications
| TNFAIP2 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP2 | |
| B94; Exoc3l3; exocyst complex component 3-like 3; M-sec; primary response gene B94 protein; TNF alpha induced protein 2; TNF alpha-induced protein 2; TNF alpha-IP 2; Tnfaip2; tnfb94; Tnfip2; tumor necrosis factor alpha-induced protein 2; tumor necrosis factor induced protein 2; tumor necrosis factor, alpha induced protein 2; tumor necrosis factor, alpha-induced protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7127 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q03169 | |
| TNFAIP2 | |
| A 19 amino acid synthetic peptide near the center of human TNFAIP2 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction