Learn More
TNFAIP6, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human TNFAIP6 peptide using ARM Technology.
Supplier: Abnova Corporation H00007130K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a secretory protein that contains a hyaluronan-binding domain, and thus is a member of the hyaluronan-binding protein family. The hyaluronan-binding domain is known to be involved in extracellular matrix stability and cell migration. This protein has been shown to form a stable complex with inter-alpha-inhibitor (I alpha I), and thus enhance the serine protease inhibitory activity of I alpha I, which is important in the protease network associated with inflammation. The expression of this gene can be induced by tumor necrosis factor alpha and interleukin-1. The expression can also be induced by mechanical stimuli in vascular smooth muscle cells, and is found to be correlated with proteoglycan synthesis and aggregation. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
TNFAIP6 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
TNFAIP6 | |
TNFAIP6 | |
A synthetic peptide of human TNFAIP6 | |
RUO | |
7130 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
tumor necrosis factor, alpha-induced protein 6 | |
TNFAIP6 | |
TSG-6, TSG6 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.