Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFAIP6, Polyclonal, Invitrogen™

Catalog No. PIPA127927
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127927 50 μg
1 options

Catalog No. PIPA127927

Supplier: Thermo Scientific PA127927

Chicken Polyclonal Antibody

Possibly involved in cell-cell and cell-matrix interactions during inflammation and tumorigenesis. The protein encoded by this gene is a secretory protein that contains a hyaluronan-binding domain. and thus is a member of the hyaluronan-binding protein fa

PA1-27927 detects TNFAIP6 from human samples.PA1-27927 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Tumor necrosis factorinducible protein TSG6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFAIP6
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene TNFAIP6
Gene Accession No. P98066
Gene Alias TSG-6, TSG6
Gene Symbols TNFAIP6
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Tumor necrosis factorinducible protein TSG6.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7130
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.