Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TNFRSF14 Clone: 2G6-2C7,

Catalog No. AP1159100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AP1159100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP1159100UG Supplier MilliporeSigma™ Supplier No. AP1159100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TNFRSF14 Clone: 2G6-2C7 in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immuno Blotting

TNFRSF14 is a member of the TNF-receptor superfamily. This receptor was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to this receptor protein has been shown to be part of the viral entry mechanism. The cytoplasmic region of this receptor was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFRSF14
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2G6-2C7
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen Full-length, recombinant, human TNFRSF14 (aa 38-248), expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the TNFRSF14 protein in human pancreatic cancer tissue.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.