Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RELT tumor necrosis factor receptor, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004750 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-750 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-750 Supplier Abnova Corporation Supplier No. H00084957A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TNFRSF19L.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is especially abundant in hematologic tissues. It has been shown to activate the NF-kappaB pathway and selectively bind TNF receptor-associated factor 1 (TRAF1). This receptor is capable of stimulating T-cell proliferation in the presence of CD3 signaling, which suggests its regulatory role in immune response. Two alternatively spliced transcript variants of this gene encoding the same protein have been reported. [provided by RefSeq

Sequence: STTLWQCPPGEEPDLDPGQGTLCRPCPPGTFSAAWGSSPCQPHARCSLWRRLEAQVGMATRDTLCGDCWPGWFGPWGVPRVPCQPCSWAPLGTHGCDEW

Specifications

Antigen RELT tumor necrosis factor receptor
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TNFRSF19L.
Formulation 50% glycerol
Gene RELT
Gene Accession No. NM_032871
Gene Alias FLJ14993/TNFRSF19L
Gene Symbols RELT
Host Species Mouse
Immunogen TNFRSF19L (NP_116260, 26 a.a. ∼ 124 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Membrane Receptors
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84957
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.