Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-TNFSF15, Clone: 4F3, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation H00009966M05
Specifications
TNFSF15 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC074941.2 | |
TNFSF15 | |
TNFSF15 (AAH74941.1, 72 a.a. ∼ 251 a.a) partial recombinant protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
4F3 | |
Mouse monoclonal antibody raised against a partial recombinant TNFSF15. | |
TNFSF15 | |
MGC129934/MGC129935/TL1/TL1A/VEGI/VEGI192A | |
Mouse | |
Affinity chromatography | |
RUO | |
9966 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction