Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

troponin T type 1 (skeletal, slow), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016919 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-919 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-919 Supplier Abnova Corporation Supplier No. H00007138B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TNNT1 protein.

This gene encodes a protein that is a subunit of troponin, which is a regulatory complex located on the thin filament of the sarcomere. This complex regulates striated muscle contraction in response to fluctuations in intracellular calcium concentration. This complex is composed of three subunits: troponin C, which binds calcium, troponin T, which binds tropomyosin, and troponin I, which is an inhibitory subunit. This protein is the slow skeletal troponin T subunit. Mutations in this gene cause nemaline myopathy type 5, also known as Amish nemaline myopathy, a neuromuscular disorder characterized by muscle weakness and rod-shaped, or nemaline, inclusions in skeletal muscle fibers which affects infants, resulting in death due to respiratory insufficiency, usually in the second year. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MSDTEEQEYEEEQPEEEAAEEEEEEEERPKPSRPVVPPLIPPKIPEGERVDFDDIHRKRMEKDLLELQTLIDVHFEQRKKEEEELVALKERIERRRSERAEQQRFRTEKERERQAKLAEEKMRKEEEEAKKRAEDDAKKKKVLSNMGAHFGGYLVKAEQKRGKRQTGREMKVRILSERKKPLDIDYMGEEQLREKAQELSDWIHQLESEKFDLMAKLKQQKYEINVLYNRISHAQKFRKGAGKGRVGGRWK

Specifications

Antigen troponin T type 1 (skeletal, slow)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TNNT1 protein.
Formulation PBS with no preservative; pH 7.4
Gene TNNT1
Gene Accession No. NM_003283
Gene Alias ANM/FLJ98147/MGC104241/STNT/TNT/TNTS
Gene Symbols TNNT1
Host Species Mouse
Immunogen TNNT1 (NP_003274, 1 a.a. ∼ 251 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7138
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.