Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNP Mouse anti-TNP, Unlabeled, Clone: 107.3, BD

Catalog No. BDB554054 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554054 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554054 Supplier BD Biosciences Supplier No. 554054
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 107.3 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human or mouse cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone 107.3 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 107.3
Isotype: IgG1 κ
Immunogen: TNP-keyhole limpet hemocyanin

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNP
Applications ELISA
Classification Monoclonal
Clone 107.3
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen TNP-keyhole limpet hemocyanin
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1
Target Molecule TNP
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.