Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transducer of ERBB2, 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016992 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-992 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-992 Supplier Abnova Corporation Supplier No. H00010140B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TOB1 protein.

This gene encodes a member of the tob/btg1 family of anti-proliferative proteins that have the potential to regulate cell growth. When exogenously expressed, this protein supresses cell growth in tissue culture. The protein undergoes phophorylation by a serine/threonine kinase, 90kDa ribosomal S6 kinase. Interactions of this protein with the v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 gene product p185 interferes with growth suppression. This protein inhibits T cell proliferation and transcription of cytokines and cyclins. The protein interacts with both mothers against decapentaplegic Drosophila homolog 2 and 4 to enhance their DNA binding activity. This interaction inhibits interleukin 2 transcription in T cells. [provided by RefSeq

Sequence: MQLEIQVALNFIISYLYNKLPRRRVNIFGEELERLLKKKYEGHWYPEKPYKGSGFRCIHIGEKVDPVIEQASKESGLDIDDVRGNLPQDLSVWIDPFEVSYQIGEKGPVKVLYVDDNNENGCELDKEIKNSFNPEAQVFMPISDPASSVSSSPSPPFGHSAAVSPTFMPRSTQPLTFTTATFAATKFGSTKMKNSGRSNKVARTSPINLGLNVNDLLKQKAISSSMHSLYGLGLGSQQQPQQQQQPAQPPPPPPPPQQQQQQKTSALSPNAKEFIFPNMQGQGSSTNGMFPGDSPLNLSPLQYSNAFDVFAAYGGLNEKSFVDGLNFSLNNMQYSNQQFQPVMAN

Specifications

Antigen transducer of ERBB2, 1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TOB1 protein.
Formulation PBS with no preservative; pH 7.4
Gene TOB1
Gene Accession No. NM_005749
Gene Alias APRO6/MGC104792/MGC34446/PIG49/TOB/TROB/TROB1
Gene Symbols TOB1
Host Species Mouse
Immunogen TOB1 (NP_005740.1, 1 a.a. ∼ 345 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 10140
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.