Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOB2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen TOB2
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-1.0 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224619SS
SDP
View Documents
Novus Biologicals
NBP224619SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224619
SDP
View Documents
Novus Biologicals
NBP224619
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

TOB2 Polyclonal specifically detects TOB2 in Human, Mouse, Bovine, Canine, Chicken, Equine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

TOB2
Western Blot 0.5-1.0 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Bovine, Canine, Chicken, Equine, Primate
Q14106
10766
A portion of amino acids 50-100 of human TOB2 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Cell Cycle and Replication
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
bK223H9, protein Tob2, Protein Tob4, TOB4KIAA1663, TOBL, Transducer of erbB-2 2, transducer of ERBB2, 2, TROB2
TOB2
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, mouse and rat, 94% homologous in cow, and 88% homologous in horse and dog.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.