Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 (CD284) Mouse anti-Human, PE, Clone: TF901, BD

Catalog No. BDB564215 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564215 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564215 Supplier BD Biosciences Supplier No. 564215
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TF901 monoclonal antibody specifically binds to TLR4 (Toll-like receptor 4) which is also known as CD284. TLR4 is a 110 kDa type I transmembrane glycoprotein that belongs to the Toll-like receptor (TLR) family. TLR4 is expressed on monocytes, macrophages, granulocytes, dendritic cells and endothelial cells. TLR4 binds lipopolysaccharide (LPS) which is a major membrane component of Gram-negative bacteria. It thus acts as an innate immune recognition receptor against many pathogens. In association with MD-2, LPS binds to and signals through TLR4 receptors by MyD88-dependent and MyD88-independent pathways to induce the cellular production of proinflammatory cytokines. TF901 antibody binding is not influenced by the human TLR4 D299G/T399I polymorphism. Preincubation of TLR4-positive cells with the TF901 antibody blocks the subsequent binding of the HTA125 antibody which is also specific for human TLR4.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR4 (CD284)
Applications Flow Cytometry
Classification Monoclonal
Clone TF901
Conjugate PE
Description TLR4; CD284; Toll-like Receptor 4; ARMD10
Host Species Mouse
Immunogen Human TLR4 Transfected Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.