Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tom20 Mouse, Unlabeled, Clone: 29, BD

Catalog No. BDB612278 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612278 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612278 Supplier BD Biosciences Supplier No. 612278
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Many mitochondrial proteins are encoded by nuclear genes and synthesized in the cytosol as pre-proteins. These pre-proteins are targeted to the mitochondria where they are translocated across the outer and inner membranes of the mitochondria. At the outer membrane of the mitochondria, this process requires a multiprotein complex called the Translocase of the Outer Membrane of the mitochondria (TOM). The TOM complex includes receptor components, Tom20, 22, 37, and 70, as well as a multisubunit general import pore complex of 400kDa. Tom20 contains a hydrophobic N-terminal region that anchors it to the outer membrane of the mitochondria, and a cytosolic region that contains a domain that facilitates binding to the basic amphiphilic targeting sequences of pre-proteins and a tetratricopeptide repeat (TPR) motif involved in interactions with Tom70. In yeast, Tom20 and Tom22 mediate the import of all pre-proteins known to use the general import machinery of the mitochondria. Thus, Tom20 is an important receptor for many mitochondrial pre-proteins that use the outer mitochondrial translocation complex.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tom20
Applications Western Blot
Classification Monoclonal
Clone 29
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Tom20 aa. 47-145
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.