Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

translocase of outer mitochondrial membrane 20 homolog (yeast), Mouse, Clone: 4F3, Abnova™

Catalog No. 89014244 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-244 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-244 Supplier Abnova Corporation Supplier No. H00009804M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TOMM20.

Sequence: MVGRNSAIAAGVCGALFIGYCIYFDRKRRSDPNFKNRLRERRKKQKLAKERAGLSKLPDLKDAEAVQKFFLEEIQLGEELLAQGEYEKGVDHLTNAIAVCGQPQQLLQVLQQTLPPPVFQMLLTKLPTISQRIVSAQSLAEDDVE

Specifications

Antigen translocase of outer mitochondrial membrane 20 homolog (yeast)
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 4F3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant TOMM20.
Formulation PBS with no preservative; pH 7.4
Gene TOMM20
Gene Accession No. BC066335
Gene Alias KIAA0016/MAS20/MGC117367/MOM19/TOM20
Gene Symbols TOMM20
Host Species Mouse
Immunogen TOMM20 (AAH66335, 1 a.a. ∼ 145 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9804
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.