Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TOMM20 Clone: 4F3,

Catalog No. ST1705100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1705100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1705100UG Supplier MilliporeSigma™ Supplier No. ST1705100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TOMM20 Clone: 4F3 in Human, Mouse, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immuno Blotting

The TOMM20 protein is a central component of the receptor complex responsible for the recognition and translocation of cytosolically synthesized mitochondrial preproteins. Together with TOM22 , it functions as the transit peptide receptor at the surface of the mitochondrion outer membrane and facilitates the movement of preproteins into the TOM40 translocation pore.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM20
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4F3
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen Full-length, recombinant, human TOMM20 (aa 1-146), expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽12-16kDa TOMM20 protein in HeLa, PC12, and NIH3T3 cells and human small intestine tissue.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.