Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TOMM22 Clone: 4G4,

Catalog No. ST1704100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1704100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1704100UG Supplier MilliporeSigma™ Supplier No. ST1704100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TOMM22 Clone: 4G4 in Human, Mouse, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immuno Blotting

The protein encoded by this gene is an integral membrane protein of the mitochondrial outer membrane. The encoded protein interacts with TOMM20 and TOMM40, and forms a complex with several other proteins to import cytosolic preproteins into the mitochondrion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM22
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4G4
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen Full-length, recombinant, human TOMM22 (aa 1-143), expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽18kDa TOMM22 protein in PC-12, NIH3T3, Raw 264.7, and HeLa cells and human small intestine tissue.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.