Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tomosyn Mouse, Unlabeled, Clone: 15, BD

Catalog No. BDB611296 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611296 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611296 Supplier BD Biosciences Supplier No. 611296

Mouse Monoclonal Antibody

Neuronal signal transmission and neurotransmitter release from the presynaptic nerve terminal is mediated by the synaptic vesicle cycle. Syntaxin plays a central role during vesicle docking and fusion through interactions with many vesicle components. During docking, vSNAREs (VAMP/synaptobrevin, synaptotagmin) on the synaptic vesicle and tSNAREs (SNAP-25, syntaxin) on the plasma membrane interact to form a 7S complex, which is essential to docking and fusion. Syntaxin associates with munc18/n-sec1 prior to and/or during the
formation of the 7S complex. This interaction may inhibit syntaxin binding proteins (VAMP, SNAP-25) that facilitate vesicle docking or fusion. Tomosyn, a syntaxin binding protein, displaces munc18 from syntaxin-1 and forms a novel 10S complex with syntaxin-1, SNAP-25 and synaptogamin. There are two splice variants of tomosyn designated b-tomosyn and s-tomosyn, while the original is referred to as m-tomosyn. Although b-tomosyn is ubiquitously expressed, s-tomosyn and m-tomosyn are expressed primarily in brain.

Host Species: Mouse
Clone: 15
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat Tomosyn aa. 910-1105
Formula Weight [Chemical]: 130kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tomosyn
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Tomosyn aa. 910-1105
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.