Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

topoisomerase (DNA) III beta, Mouse, Clone: 1H8, Abnova™

Catalog No. 89001109 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-109 Supplier Abnova Corporation Supplier No. H00008940M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TOP3B.

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus relaxing the supercoils and altering the topology of DNA. The enzyme interacts with DNA helicase SGS1 and plays a role in DNA recombination, cellular aging and maintenance of genome stability. Alternative splicing of the C-terminus of this gene results in three transcript variants which have distinct tissue specificity; however, not all variants have been fully described. [provided by RefSeq

Sequence: MKTVLMVAEKPSLAQSIAKILSRGSLSSHKGLNGACSVHEYTGTFAGQPVRFKMTSVCGHVMTLDFLGKYNKWDKVDPAELFSQAPTEKKEANPKLNMVKFLQVEGRGCDYIVLWLDCDKEGENICFEVLDAVLPVMNKAHGGEKTVFRARFSSITDTDICNAMACLGEPDHNEALSVDARQELDLRIGCAFTRFQTKYFQGKYGDLDSSLISFGPCQTPTLGFCVERHDKIQSFKPETYWVLQAKVNTDKDRSLLLDWDRVRVFDREIAQMFLNMTKLEKEAQVEATSRKEKAKQRPLALNTVEMLRVASSSLGMGPQHAMQTAERLYTQGYISYPRTETTHYPENFDLKGSLRQQANHPYWANTVKRLLAEGINRPRKGHDAGDHPPITPMKSATEAELGGDAWRLYEYITRHFIATVSHDCKYLQSTISFRIGPELFTCSGKTVLSPGFTEVMPWQSVPLEESLPTCQRGDAFPVGEVKMLEKQTNPPDYLTEAELITLMEKHGIGTDASIPVHINNICQRNYVTVESGRRLKPTNLGIVLVHGYYKIDAELVLPTIRSAVEKQLNLIAQGKADYRQVLGHTLDVFKRKFHYFVDSIAGMDELMEVSFSPLAATGKPLSRCGKCHRFMKYIQAKPSRLHCSHCDETYTLPQNGTIKLYKELRCPLDDFELVLWSSGSRGKSYPLCPYCYNHPPFRDMKKGMGCNECTHPSCQHSLSMLGIGQCVECESGVLVLDPTSGPKWKVACNKCNVVAHCFENAHRVRVSADTCSVCEAALLDVDFNKAKSPLPGDETQHMGCVFCDPVFQELVELKHAASCHPMHRGGPGRRQGRGRGRARRPPGKPNPRRPKDKMSALAAYFV

Specifications

Antigen topoisomerase (DNA) III beta
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1H8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant TOP3B.
Formulation PBS with no preservative; pH 7.4
Gene TOP3B
Gene Accession No. BC002432
Gene Alias FLJ39376
Gene Symbols TOP3B
Host Species Mouse
Immunogen TOP3B (AAH02432, 1 a.a. ∼ 862 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8940
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.