Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Topoisomerase II Mouse anti-Human, Clone: KiS1, MilliporeSigma™

Catalog No. MAB4197MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4197MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4197MI Supplier MilliporeSigma Supplier No. MAB4197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Topoisomerase II Antibody, clone KiS1 is a Mouse Monoclonal Antibody for detection of Topoisomerase II also known as Ki-S1 and has been validated in flow cytometry, western blot, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Topoisomerase II
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone KiS1
Conjugate Unconjugated
Formulation Purified immunoglobulin.
Gene Accession No. NM_001067.2
Gene Symbols TOP2A, TOP2, TP2A, EC 5.99.1.3
Host Species Mouse
Immunogen Nuclear protein preparation from the human cell line U937.50
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.