Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Topoisomerase II Clone: SWT3D1,

Catalog No. NA14100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NA14100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NA14100UG Supplier MilliporeSigma™ Supplier No. NA14100UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Specifically detects Topoisomerase II Clone: SWT3D1 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

Topoisomerase II is a nuclear enzyme whose primary function is to induce temporary double-strand breaks in DNA and to facilitate the passage of the duplex DNA segments through these breaks. Phosphorylation of topoisomerase II in a cell cycle regulated manner appears to be important for progression through the cell cycle. The enzyme exists as two discrete isozymes, topoisomerase IIα (∽170kDa) and topoisomerase IIβ (∽180kDa) which are under independent transcriptional regulation. Both isozymes have been identified in human and mouse cells but only the topoisomerase IIα form appears in yeast. The gene encoding topoisomerase IIα has been mapped to chromosome 17 and is in close proximity to the gene for erbB-2. Altered expression of topoisomerase II has been documented in many cancers including AML, CLL, small cell lung cancer, breast and ovarian cancer, and colon cancer. Based on this observation, topoisomerase II has been identified as the target of many widely used antineoplastic drugs as well as the target of rational drug design.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Topoisomerase II
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SWT3D1
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 2% gelatin.
Host Species Mouse
Immunogen recombinant human topoisomerase IIα
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes ∽170kDa topoisomerase II protein in HeLa cells and breast carcinoma tissue.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.