Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TORC2 (454-607), Polyclonal

Catalog No. ST109950UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST109950UL 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST109950UL Supplier MilliporeSigma™ Supplier No. ST109950UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects TORC2 (454-607) Clone: in Human, Mouse, Rat samples, and it is validated for Immunoprecipitation

TORCs (Transducers Of Regulated cAMP Response Element-Binding (CREB) protein) are members of a conserved family of co-activators that enhance CRE-dependent transcription by a phosphorylation-independent interaction with the bZIP DNA binding/dimerization domain of CREB. Studies suggest that TORC phosphorylation and nuclear/cytoplasmic shuttling play an important role in the regulation of gluconeogenesis by cAMP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TORC2 (454-607)
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen a recombinant protein consisting of amino acids 454-607 of mouse TORC2 fused to GST
Purification Method Unpurified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽79-85kDa TORC2 protein in primary hepatocytes, HepG2, NIH3T3, Min6, and FAO cells.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.