Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TorsinA Mouse anti-Human, Mouse, Rat, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5550
Description
This Anti-TorsinA Antibody is validated for use in immunoprecipitation, western blot, IC for the detection of TorsinA.
Specifications
| TorsinA | |
| Monoclonal | |
| Liquid. Contaings no preservative | |
| TOR1A, DQ1, DYT1, TOR1B, DQ2 | |
| Maltose binding protein fusion protein from human torsinA. | |
| 100 μL | |
| Human, Mouse, Rat | |
| Ascites |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| NM_000113.2 | |
| Mouse | |
| Ascites | |
| Primary | |
| −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction