Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPO, Mouse, Clone: 6G12, Abnova™

Catalog No. 89024826 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-826 Supplier Abnova Corporation Supplier No. MAB8343

Mouse monoclonal antibody raised against TPO.

This gene encodes a membrane-bound glycoprotein. The encoded protein acts as an enzyme and plays a central role in thyroid gland function. The protein functions in the iodination of tyrosine residues in thyroglobulin and phenoxy-ester formation between pairs of iodinated tyrosines to generate the thyroid hormones, thyroxine and triiodothyronine. Mutations in this gene are associated with several disorders of thyroid hormonogenesis, including congenital hypothyroidism, congenital goiter, and thyroid hormone organification defect IIA. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Additional splice variants have been described but their biological natures have not been determined. [provided by RefSeq

Specifications

Antigen TPO
Applications ELISA
Classification Monoclonal
Clone 6G12
Conjugate Unconjugated
Description thyroid peroxidase
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene TPO
Gene Alias MSA, TPX
Gene Symbols TPO
Host Species Mouse
Immunogen Purified human TPO.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7173
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.