Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPOR/c-Mpl Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 06944MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-944-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-944-MI Supplier MilliporeSigma Supplier No. 06944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TPOR/c-Mpl Antibody is an antibody against TPOR/c-Mpl for use in immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPOR/c-Mpl
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols MPL; TPOR; MPLV; CD110; TPO-R; C-MPL; C-mpl
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 615-628 of human TPO (thrombopoietin) receptor (C-ANHSYLPLSYWQQP)
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005373
Target Species Human, Mouse
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.