Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA-1-60 Mouse anti-Human, Clone: TRA-1-60, MilliporeSigma™

Catalog No. MAB4360MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4360MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4360MI Supplier MilliporeSigma Supplier No. MAB4360
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-TRA-1-60 Antibody, clone TRA-1-60 is validated for use in western blot, flow cytometry, immunofluoresence, immunoprecipitation, IC for the detection of TRA-1-60.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-60
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TRA-1-60
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgM liquid in 0.05M Potassium Phosphate, 0.3M NaCl pH 8.0 with 0.05% Sodium Azide.
Gene Symbols TRA-1-60
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.