Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA-1-60, Mouse, Clone: TRA-1-60(R), Abnova™

Catalog No. 89024822 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-822 Supplier Abnova Corporation Supplier No. MAB2391
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native TRA-1-60.

Specifications

Antigen TRA-1-60
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TRA-1-60(R)
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native TRA-1-60.
Dilution Flow Cytometry (1:50-1:200) Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Host Species Mouse
Immunogen Native purified from human embryonal carcinoma cell line 2102Ep.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity TRA-1-60 recognizes antigens that are associated with a pericellular matrix proteoglycan. TRA-1-60 reacts with a sialidase-sensitive epitope.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.