Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA-1-85 Mouse anti-Human, Unlabeled, Clone: TRA-1-85, BD

Catalog No. BDB563020 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563020 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563020 Supplier BD Biosciences Supplier No. 563020
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TRA-1-85 monoclonal antibody (mAb) specifically binds to the high frequency blood group antigen OKa. The OKa blood group antigen is a 35-69 kDa glycoprotein also known as CD147, EMMPRIN, M6, or Basigin. The OKa group is widely expressed on most human tissues including, but not limited to kidney, peripheral blood leukocytes, liver, pancreas, colon, skin, and brain. TRA-1-85 mAb is often used to discern human cells from non-human cells in multiple applications including animal transplantation studies. During development, the purified TRA-1-85 mAb was found to detect OKa blood group antigen by
western blot analysis of cellular lysates, by immunofluorescent staining of fixed cells, and by flow cytometric analysis of live cells

Host Species: Mouse
Clone: TRA-1-85
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human Embryonal Carcinoma Cell Line

Bioimaging, Flow Cytometry, Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-85
Applications Flow Cytometry
Classification Monoclonal
Clone TRA-1-85
Concentration 0.5mg/mL
Conjugate Unconjugated
Description OKa, CD147, EMMPRIN, Basigin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Embryonal Carcinoma Cell Line
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.