Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trace Amine Receptor 5 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB9497MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9497MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9497MI Supplier MilliporeSigma Supplier No. AB9497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Trace Amine Receptor 5 Antibody is validated for use in immunohistochemistry (paraffin) for the detection of Trace Amine Receptor 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trace Amine Receptor 5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide.
Gene Symbols TAAR8; TaR-5; TAR5; TRAR5; TA5; GPR102
Host Species Rabbit
Immunogen Synthetic peptide from the 3rd cytoplasmic domain of human Trace Amine Receptor 5.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_053278.1
Target Species Human
Content And Storage −70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.