Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TRADD Clone: 1G6.1,

Catalog No. MABC988MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC988MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC988MI Supplier MilliporeSigma™ Supplier No. MABC988
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TRADD Clone: 1G6.1 in Human, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Tumor necrosis factor receptor type 1-associated DEATH domain protein (UniProt Q15628; also known as TNFR1-associated death domain protein, TNFRSF1A-associated via death domain, Tumor necrosis factor receptor-1-associated protein) is encoded by the TRADD (also known as Hs.89862) gene (Gene ID 8717) in human. TRADD is an adapter protein that interacts with the TNFR1 receptor, and mediates the activation of programmed cell death and NF-κB activation. TRADD exhibits similar structural and functional features, as well as subcellular localization as the adaptor protein Fas-associated death domain (FADD). Both proteins contain a C-terminal death domain (DD) and function as mediators of death receptors-mediated signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRADD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G6.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q15628
Gene Symbols TRADD
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human TRADD.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_003780
Test Specificity Reactivity toward human TRADD spliced isoform 2 has not been determined.
Target Species Human, Rat
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.