Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF2 Mouse anti-Human, Unlabeled, Clone: C90-481, BD

Catalog No. BDB558890 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558890 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558890 Supplier BD Biosciences Supplier No. 558890
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAFs (TNF receptor associated factors) are signal transducing molecules which participate in forming TNF/Nerve Growth Factor receptor-associated signalling complexes. TRAF proteins share a novel C-terminal homology region, the “TRAF” domain, a coiled-coiled region and the majority have Zinc/RING finger motifs. At least six TRAF proteins (TRAFs 1-6) have been identified. These proteins are cytoplasmic adapters; they bind to cytoplasmic domains of various receptors and can function to recruit other proteins to a signaling complex thereby promoting intracellular signal transduction. Specifically, TRAF2 has been shown to interact with the following surface receptors: TNFRII, CD27, CD30, CD40, 4-1BB, Ox40, HVEM/ATAR and LMP-1. TRAF2 has also been shown to associate with intracellular proteins, including TRADD, FADD, I-TRAF/TANK, TRIP, A20, c-IAP1 and 2, Casper, RIP and NIK. Thus, the interaction of TRAF2 with these various receptors and intracellular proteins plays an important role modulating downstream events which can result in inducing cell death or keeping cells alive. For example, cells deficient in TRAF2 demonstrated increased sensitivity to TNF-induced cell death, but a decrease in TNF-induced JNK activation, and little change on NF-kB activation. TRAF2 is detected at ∽ 53kDa in western blot analysis. The C90-481 antibody reacts with human TRAF2. A synthetic protein freagment corresponding to amino acids 93-199 of human TRAF2 was used as immunogen.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF2
Applications Western Blot
Classification Monoclonal
Clone C90-481
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human TRAF2 aa. 93-199
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.