Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF3 interacting protein 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018244 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-244 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-244 Supplier Abnova Corporation Supplier No. H00010758D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TRAF3IP2 protein.

This gene encodes a protein involved in regulating responses to cytokines by members of the Rel/NF-kappaB transcription factor family. These factors play a central role in innate immunity in response to pathogens, inflammatory signals and stress. This gene product interacts with TRAF proteins (tumor necrosis factor receptor-associated factors) and either I-kappaB kinase or MAP kinase to activate either NF-kappaB or Jun kinase. Several alternative transcripts encoding different isoforms have been identified. Another transcript, which does not encode a protein and is transcribed in the opposite orientation, has been identified. Overexpression of this transcript has been shown to reduce expression of at least one of the protein encoding transcripts, suggesting it has a regulatory role in the expression of this gene. [provided by RefSeq

Sequence: MNRSIPVEVDESEPYPSQLLKPIPEYSPEEESEPPAPNIRNMAPNSLSAPTMLHNSSGDFSQAHSTLKLANHQRPVSRQVTCLRTQVLEDSEDSFCRRHPGLGKAFPSGCSAVSEPASESVVGALPAEHQFSFMEKRNQWLVSQLSAASPDTGHDSDKSDQSLPNASADSLGGSQEMVQRPQPHRNRAGLDLPTIDTGYDSQPQDVLGIRQLERPLPLTSVCYPQDLPRPLRSREFPQFEPQRYPACAQMLPPNLSPHAPWNYHYHCPGSPDHQVPYGHDYPRAAYQQVIQPALPGQPLPGASVRGLHPVQKVILNYPSPWDQEERPAQRDCSFPGLPRHQDQPHHQPPNRAGAPGESLECPAELRPQVPQPPSPAAVPRPPSNPPARGTLKTSNLPEELRKVFITYSMDTAMEVVKFVNFLLVNGFQTAIDIFEDRIRGIDIIKWMERYLRDKTVMIIVAISPKYKQDVEGAESQLDEDEHGLHTKYIHRMMQIEFIKQGSMNFRFIPVLFPNAKKEHVPTWLQNTHVYSWPKNKKNILLRLLREEEYVAPPRGPLPTLQVVPL

Specifications

Antigen TRAF3 interacting protein 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TRAF3IP2 protein.
Formulation PBS with no preservative; pH 7.4
Gene TRAF3IP2
Gene Accession No. NM_147686
Gene Alias ACT1/C6orf2/C6orf4/C6orf5/C6orf6/CIKS/DKFZp586G0522/MGC3581
Gene Symbols TRAF3IP2
Host Species Rabbit
Immunogen TRAF3IP2 (NP_679211.1, 1 a.a. ∼ 565 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10758
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.