Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAF4 Mouse, Unlabeled, Clone: 43, BD

Catalog No. BDB610866 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610866 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610866 Supplier BD Biosciences Supplier No. 610866

Mouse Monoclonal Antibody

CART1 gene, located on q11-q12 region of the chromosome 17 long arm, is commonly amplified in human breast carcinomas. The CART1 protein contains several motifs known as the RING, CART, and TRAF domains. The RING domain is cysteine rich (C3HC3D) and is located at the N-terminus. The CART domain is centrally located in the protein and is a cysteine-rich region composed of three repeats of HC3HC3. The TRAF domain is located at the C-terminus and is involved in protein-protein interactions. These domains are also found in the human CD40-binding protein and the mouse TNF receptor-associated factor 2 (TRAF2), which are involved in TNF receptor signaling pathways. CART1 is also known as TRAF4 and is a member of the TRAF family of proteins. TRAF4/CART1 is expressed in breast carcinomas and metastatic axillary lymph nodes, while the mRNA is found in malignant epithelial cells. Immunohistochemistry of a breast carcinoma revealed that TRAF4 is located within the nucleus. Therefore, TRAF4 may play a role in the development of breast carcinomas via TNF-dependent signaling pathways.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF4
Applications Western Blot
Classification Monoclonal
Clone 43
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CART1 aa. 2-160
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.