Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF receptor-associated factor 4, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016984 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-984 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-984 Supplier Abnova Corporation Supplier No. H00009618B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TRAF4 protein.

This gene encodes a member of the TNF receptor associated factor (TRAF) family. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. The encoded protein has been shown to interact with neurotrophin receptor, p75 (NTR/NTSR1), and negatively regulate NTR induced cell death and NF-kappa B activation. This protein has been found to bind to p47phox, a cytosolic regulatory factor included in a multi-protein complex known as NAD(P)H oxidase. This protein thus, is thought to be involved in the oxidative activation of MAPK8/JNK. Alternatively spliced transcript variants have been observed but the full-length nature of only one has been determined. [provided by RefSeq

Sequence: MPGFDYKFLEKPKRRLLCPLCGKPMREPVQVSTCGHRFCDTCLQEFLSEGVFKCPEDQLPLDYAKIYPDPELEVQVLGLPIRCIHSEEGCRWSGPLRHLQGHLNTCSFNVIPCPNRCPMKLSRRDLPAHLQHDCPKRRLKCEFCGCDFSGEAYESHEGMCPQESVYCENKCGARMMRRLLAQHATSECPKRTQPCTYCTKEFVFDTIQSHQYQCPRLPVACPNQCGVGTVAREDLPGHLKDSCNTALVLCPFKDSGCKHRCPKLAMARHVEESVKPHLAMMCALVSRQRQELQELRRELEELSVGSDGVLIWKIGSYGRRLQEAKAKPNLECFSPAFYTHKYGYKLQVSAFLNGNGSGEGTHLSLYIRVLPGAFDNLLEWPFARRVTFSLLDQSDPGLAKPQHVTETFHPDPNWKNFQKPGTWRGSLDESSLGFGYPKFISHQDIRKRNYVRDDAVFIRAAVELPRKILS

Specifications

Antigen TNF receptor-associated factor 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TRAF4 protein.
Formulation PBS with no preservative; pH 7.4
Gene TRAF4
Gene Accession No. NM_004295
Gene Alias CART1/MLN62/RNF83
Gene Symbols TRAF4
Host Species Mouse
Immunogen TRAF4 (NP_004286.2, 1 a.a. ∼ 470 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9618
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.