Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF receptor-associated factor 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018334 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-334 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-334 Supplier Abnova Corporation Supplier No. H00007188D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TRAF5 protein.

The scaffold protein encoded by this gene is a member of the tumor necrosis factor receptor-associated factor (TRAF) protein family and contains a meprin and TRAF homology (MATH) domain, a RING-type zinc finger, and two TRAF-type zinc fingers. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. This protein is one of the components of a multiple protein complex which binds to tumor necrosis factor (TNF) receptor cytoplasmic domains and mediates TNF-induced activation. Alternate transcriptional splice variants have been characterized. [provided by RefSeq

Sequence: MAYSEEHKGMPCGFIRQNSGNSISLDFEPSIEYQFVERLEERYKCAFCHSVLHNPHQTGCGHRFCQHCILSLRELNTVPICPVDKEVIKSQEVFKDNCCKREVLNLYVYCSNAPGCNAKVILGRYQDHLQQCLFQPVQCSNEKCREPVLRKDLKEHLSASCQFRKEKCLYCKKDVVVINLQNHEENLCPEYPVFCPNNCAKIILKTEVDEHLAVCPEAEQDCPFKHYGCAVTDKRRNLQQHEHSALREHMRLVLEKNVQLEEQISDLHKSLEQKESKIQQLAETIKKLEKEFKQFAQLFGKNGSFLPNIQVFASHIDKSAWLEAQVHQLLQMVNQQQNKFDLRPLMEAVDTVKQKITLLENNDQRLAVLEEETNKHDTHINIHKAQLSKNEERFKLLEGTCYNGKLIWKVTDYKMKKREAVDGHTVSIFSQSFYTSRCGYRLCARAYLNGDGSGRGSHLSLYFVVMRGEFDSLLQWPFRQRVTLMLLDQSGKKNIMETFKPDPNSSSFKRPDGEMNIASGCPRFVAHSVLENAKNAYIKDDTLFLKVAVDLTDLEDL

Specifications

Antigen TNF receptor-associated factor 5
Applications In situ PLA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TRAF5 protein.
Formulation PBS with no preservative; pH 7.4
Gene TRAF5
Gene Accession No. NM_001033910
Gene Alias MGC:39780/RNF84
Gene Symbols TRAF5
Host Species Rabbit
Immunogen TRAF5 (NP_001029082.1, 1 a.a. ∼ 557 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 7188
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.